t-Boc-N-Amido-PEG9-Amine
Leverantör: Apollo Scientific
Synonyms: tert-Butyl (29-amino-3,6,9,12,15,18,21,24,27-nonaoxanonacosyl)carbamate, Boc-amido-PEG9-amine
BIPG1816-100MG BIPG1816-500MG BIPG1816-1G
APOSBIPG1816-100MGEA 1125 SEK
APOSBIPG1816-100MG APOSBIPG1816-500MG APOSBIPG1816-1G
t-Boc-N-Amido-PEG9-Amine
t-Boc-N-Amido-PEG9-Amine
Formel: C₂₅H₅₂N₂O₁₁
MW: 556,687 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD03453249
CAS nummer: 890091-43-7