t-Boc-N-Amido-PEG2-acid
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatetradecan-14-oic acid, 3-[2-(2-([(tert-Butoxy)carbonyl]amino)ethoxy)ethoxy]propanoic acid, Boc-NH-PEG2-CH2CH2COOH, N-Boc-3-[2-(2-aminoethoxy)ethoxy]propionic Acid
BIPG1800-500MG
BIPG1800-100MG
BIPG1800-1G
APOSBIPG1800-500MGEA
2250
SEK
APOSBIPG1800-500MG
APOSBIPG1800-100MG
APOSBIPG1800-1G
t-Boc-N-Amido-PEG2-acid
t-Boc-N-Amido-PEG2-acid
Formel:
C₁₂H₂₃NO₆ MW: 277,314 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD22056304 CAS nummer: 1365655-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...