Fmoc-N-amido-PEG1-acid
Leverantör: Apollo Scientific
Synonyms: 3-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethoxy)propanoic acid, Fmoc-NH-PEG1-C2-acid
BIPG1395-1G BIPG1395-100MG
APOSBIPG1395-1GEA 855 SEK
APOSBIPG1395-1G APOSBIPG1395-100MG
Fmoc-N-amido-PEG1-acid
Fmoc-N-amido-PEG1-acid
Formel: C₂₀H₂₁NO₅
Storage Temperature: Omgivande
MDL Number: MFCD16294357
CAS nummer: 1654740-73-4