Bromo-PEG3-t-butyl ester
Leverantör: Apollo Scientific
Varning
Synonyms: Br-PEG3-C2-Boc
BIPG1349-250MG BIPG1349-50MG BIPG1349-100MG
APOSBIPG1349-250MGEA 1350 SEK
APOSBIPG1349-250MG APOSBIPG1349-50MG APOSBIPG1349-100MG
Bromo-PEG3-t-butyl ester
Bromo-PEG3-t-butyl ester
Formel: C₁₃H₂₅BrO₅
MW: 341,239 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22574821
CAS nummer: 782475-37-0