Azido-PEG3-acid
Leverantör: Apollo Scientific
Varning
BIPG1105-250MG BIPG1105-100MG BIPG1105-500MG
APOSBIPG1105-250MGEA 2925 SEK
APOSBIPG1105-250MG APOSBIPG1105-100MG APOSBIPG1105-500MG
Azido-PEG3-acid
Azido-PEG3-acid
Formel: C₉H₁₇N₃O₅
MW: 247,248 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD23726617
CAS nummer: 1056024-94-2