Amino-PEG2-acid
Leverantör: Apollo Scientific
Varning
Synonyms: Amino-PEG2-C2-acid, 3-(2-(2-Aminoethoxy)ethoxy)propanoic acid
BIPG1031-100MG BIPG1031-250MG BIPG1031-1G
APOSBIPG1031-100MGEA 855 SEK
APOSBIPG1031-100MG APOSBIPG1031-250MG APOSBIPG1031-1G
Amino-PEG2-acid
Amino-PEG2-acid
Formel: C₇H₁₅NO₄
MW: 177,198 g/mol
Storage Temperature: Frys
MDL Number: MFCD20926377
CAS nummer: 791028-27-8