5(6)-Iodoacetamidotetramethylrhodamine

Leverantör: Apollo Scientific
Varning

Synonyms: IATR

BII7024-10MG BII7024-5MG
APOSBII7024-10MGEA 2575 SEK
APOSBII7024-10MG APOSBII7024-5MG
5(6)-Iodoacetamidotetramethylrhodamine
5(6)-Iodoacetamidotetramethylrhodamine
Formel: C₂₆H₂₄IN₃O₄
MW: 569.391 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00269791
CAS nummer: 136538-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR