6-(y,y-Dimethylallylamino)purine (2IP)
Leverantör: Apollo Scientific
|
Varning
|
BID525-1G
BID525-500MG
BID525-5G
APOSBID525-1GEA
1625
SEK
APOSBID525-1G
APOSBID525-500MG
APOSBID525-5G
6-(y,y-Dimethylallylamino)purine (2IP)
6-(y,y-Dimethylallylamino)purine (2IP)
Plant cell culture tested
Formel:
C₁₀H₁₃N₅ MW: 203,244 g/mol Smältpunkt: 190…210 °C Storage Temperature: Frys |
MDL Number:
MFCD00132998 CAS nummer: 2365-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...