D-bishomopropargylglycine

Leverantör: Apollo Scientific
Varning

BICR120-1G BICR120-200MG BICR120-500MG
APOSBICR120-1GEA 15450 SEK
APOSBICR120-1G APOSBICR120-200MG APOSBICR120-500MG
D-bishomopropargylglycine
D-bishomopropargylglycine
Formel: C₇H₁₁NO₂
Storage Temperature: Frys
MDL Number: MFCD11052908
CAS nummer: 211054-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR