L-bishomopropargylglycine

Leverantör: Apollo Scientific
Varning

BICR115-1G BICR115-200MG BICR115-500MG
APOSBICR115-1GEA 18450 SEK
APOSBICR115-1G APOSBICR115-200MG APOSBICR115-500MG
L-bishomopropargylglycine
L-bishomopropargylglycine
Formel: C₇H₁₁NO₂
Storage Temperature: Frys
MDL Number: MFCD11052904
CAS nummer: 835627-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR