2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester ≥98%
Leverantör: Apollo Scientific
Varning
Synonyms: (2S,3S,4S,5R,6R)-2-(Methoxycarbonyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4,5-triyl tris(2-methylpropanoate), Methyl 2,3,4-tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronate
BICL4368-100MG BICL4368-250MG BICL4368-500MG
APOSBICL4368-100MGEA 4275 SEK
APOSBICL4368-100MG APOSBICL4368-250MG APOSBICL4368-500MG
2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester ≥98%
2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-α-D-glucopyranuronic acid methyl ester
Formel: C₂₁H₃₀Cl₃NO₁₀
MW: 562,823 g/mol
Smältpunkt: 82…84 °C
Storage Temperature: Frys
MDL Number: MFCD09817649
CAS nummer: 150607-96-8