4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%
Leverantör: Apollo Scientific
Synonyms: Thiocresyl 2,3-di-O-acetyl-4,6-O-benzylidene-β-D-glucopyranoside, Tolyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside
BICL4353-1G BICL4353-5G BICL4353-2G
APOSBICL4353-1GEA 4275 SEK
APOSBICL4353-1G APOSBICL4353-5G APOSBICL4353-2G
4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 2,3-di-O-acetyl-4,6-O-benzylidene-1-thio-β-D-glucopyranoside
Formel: C₂₄H₂₆O₇S
MW: 458,524 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28397803
CAS nummer: 219518-20-4