4-Methylphenyl 2,3,6-tri-O-acetyl-1-thio-β-D-glucopyranoside ≥98%
Leverantör: Apollo Scientific
Synonyms: Tolyl 2,3,6-tri-O-acetyl-1-thio-β-D-glucopyranoside, Thiocresyl 2,3,6-tri-O-acetyl-β-D-glucopyranoside
BICL4351-1G BICL4351-2G BICL4351-500MG
APOSBICL4351-1GEA 8400 SEK
APOSBICL4351-1G APOSBICL4351-2G APOSBICL4351-500MG
4-Methylphenyl 2,3,6-tri-O-acetyl-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 2,3,6-tri-O-acetyl-1-thio-β-D-glucopyranoside