3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose ≥98%
Leverantör: Apollo Scientific
Synonyms: 1,2:5,6-Di-O-isopropylidene-3-O-2-propen-1-yl-α-D-glucofuranose
BICL4319-10G BICL4319-25G BICL4319-50G
APOSBICL4319-10GEA 4975 SEK
APOSBICL4319-10G APOSBICL4319-25G APOSBICL4319-50G
3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose ≥98%
3-O-Allyl-1,2:5,6-di-O-isopropylidene-α-D-glucofuranose
Formel: C₁₅H₂₄O₆
MW: 300.347 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01725444
CAS nummer: 20316-77-2