2-Chloro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside 95%
Leverantör: Apollo Scientific
BICL4310-10MG BICL4310-25MG BICL4310-5MG
APOSBICL4310-10MGEA 5775 SEK
APOSBICL4310-10MG APOSBICL4310-25MG APOSBICL4310-5MG
2-Chloro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside 95%
2-Chloro-4-nitrophenyl 2-azido-2-deoxy-β-D-galactopyranoside