4-Methylphenyl 2,3-di-O-benzyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside ≥98%
Leverantör: Apollo Scientific
Synonyms: Thiocresyl 2,3-di-O-benzy-4,6-O-benzylideneylidene-α-D-mannopyranoside, Tolyl 2,3-di-O-benzy-4,6-O-benzylidene-1-thio-α-D-mannopyranoside
BICL4304-1G BICL4304-250MG BICL4304-500MG
APOSBICL4304-1GEA 14200 SEK
APOSBICL4304-1G APOSBICL4304-250MG APOSBICL4304-500MG
4-Methylphenyl 2,3-di-O-benzyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside ≥98%
4-Methylphenyl 2,3-di-O-benzyl-4,6-O-benzylidene-1-thio-α-D-mannopyranoside
Formel: C₃₄H₃₄O₅S
MW: 554,696 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD15145129
CAS nummer: 922523-12-4