3-O-(Carboxymethyl)-D-glucose ≥95%

Leverantör: Apollo Scientific

BICL4290-5MG BICL4290-1MG BICL4290-2MG
APOSBICL4290-5MGEA 9075 SEK
APOSBICL4290-5MG APOSBICL4290-1MG APOSBICL4290-2MG
3-O-(Carboxymethyl)-D-glucose ≥95%
3-O-(Carboxymethyl)-D-glucose
Formel: C₈H₁₄O₈
MW: 238,192 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28967907
CAS nummer: 95350-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR