2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-α-D-galactopyranoside ≥99%
Leverantör: Apollo Scientific
Synonyms: 2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl trichloroacetimidate
BICL4217-1G BICL4217-2G BICL4217-5G
APOSBICL4217-1GEA 4475 SEK
APOSBICL4217-1G APOSBICL4217-2G APOSBICL4217-5G
2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-α-D-galactopyranoside ≥99%
2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-α-D-galactopyranoside
Formel: C₁₆H₂₀Cl₃NO₁₀
MW: 492,69 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07369652
CAS nummer: 353264-42-3