2-(Dimethoxymethyl)furan ≥99%

Leverantör: Apollo Scientific

Synonyms: 2-Furaldehyde dimethyl acetal

BICL4090-1G BICL4090-2G
APOSBICL4090-1GEA 4950 SEK
APOSBICL4090-1G APOSBICL4090-2G
2-(Dimethoxymethyl)furan ≥99%
2-(Dimethoxymethyl)furan
Storage Temperature: Omgivande
CAS nummer: 1453-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR