Benzyl 2,3:5,6-di-O-isopropylidene-α-D-mannofuranoside ≥98%
Leverantör: Apollo Scientific
BICL4050-10G BICL4050-2G BICL4050-5G
APOSBICL4050-10GEA 13200 SEK
APOSBICL4050-10G APOSBICL4050-2G APOSBICL4050-5G
Benzyl 2,3:5,6-di-O-isopropylidene-α-D-mannofuranoside ≥98%
Benzyl 2,3:5,6-di-O-isopropylidene-α-D-mannofuranoside
Storage Temperature: Omgivande
CAS nummer: 20689-02-5