Biochanin A-7-O-β-D-glucuronide ≥98.8%
Leverantör: Apollo Scientific
Varning
Synonyms: 5,7-Dihydroxy-4’-methoxyisoflavone-7-O-β-D-glucuronide
BICL4037-10MG BICL4037-25MG BICL4037-5MG
APOSBICL4037-10MGEA 7725 SEK
APOSBICL4037-10MG APOSBICL4037-25MG APOSBICL4037-5MG
Biochanin A-7-O-β-D-glucuronide ≥98.8%
Biochanin A-7-O-β-D-glucuronide
Storage Temperature: Omgivande
CAS nummer: 38482-76-7