Sesquicillin A
Leverantör: Apollo Scientific
|
Fara
|
BIBR1155-1MG
BIBR1155-5MG
APOSBIBR1155-1MGEA
3350
SEK
APOSBIBR1155-1MG
APOSBIBR1155-5MG
Sesquicillin A
Sesquicillin A
From microbial source
Formel:
C₂₉H₄₂O₅ Storage Temperature: Frys |
MDL Number:
MFCD08274594 CAS nummer: 51103-58-3 UN: 2811 ADR: 6.1,III |