Monocerin
Leverantör: Apollo Scientific
|
Varning
|
BIBR1065-1MG
BIBR1065-5MG
APOSBIBR1065-1MGEA
2750
SEK
APOSBIBR1065-1MG
APOSBIBR1065-5MG
Monocerin
Monocerin
Formel:
C₁₆H₂₀O₆ Storage Temperature: Omgivande |
MDL Number:
MFCD08274586 CAS nummer: 30270-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...