N-Methylindolyl-β-D-galactopyranoside monohydrate

Leverantör: Apollo Scientific
Varning

Synonyms: Green β-D-gal

BIB6109-100MG BIB6109-50MG BIB6109-250MG
APOSBIB6109-100MGEA 2325 SEK
APOSBIB6109-100MG APOSBIB6109-50MG APOSBIB6109-250MG
N-Methylindolyl-β-D-galactopyranoside monohydrate
N-Methylindolyl-β-D-galactopyranoside monohydrate
Storage Temperature: Kylskåp
MDL Number: MFCD21364772
CAS nummer: 207598-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR