Polyethylene glycol 2,000, GPR RECTAPUR®

Leverantör: VWR Chemicals

Synonyms: PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000

26608.290EA 620 SEK
26608.290 26608.360
Polyethylene glycol 2,000, GPR RECTAPUR®
Polyethylene glycol 2,000
Formel: H(OCH₂CH₂)nOH
MW: 2000 g/mol
MDL Number: MFCD00081839
CAS nummer: 25322-68-3

Order Now

Specifikation testresultat

Average molecular weight 1800 - 2500
Hydroxyl value 51 - 63
pH (5%) 4,0 - 8,0
Viscosity (20°C, 50% water) 50 - 58 mPa.s
Ignition residue max. 0.2%
Water max. 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR