Graduated narrow neck bottles, PE, with screw cap and undercap
Leverantör: KARTELL
317 319 323 324 325 326
KART317EA 7475 SEK
KART317 KART319 KART323 KART324 216-2506 KART326
Graduated narrow neck bottles, PE, with screw cap and undercap
Flaskor Flaskor, plast
These high quality graduated narrow neck bottles are made of PE (main body and under cap) and supplied with a PP cap.
  • Suitable for foodstuff contact according to EC Regulation 10/2011
  • Translucent
  • Tagging points on shoulder and cap ensures tamper-evidence by means of tags and seals
Graduated bottle for liquids and powders. Sloping shoulders reduce tendency for residue deposits. Moulded-in graduations.
Certifikat: Meets EC Regulation 10/2011. Migration Test Compliance Declaration available.