Fisetin
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3,7,3',4'-Tetrahydroxyflavone
119150010 119150050
ACRO119150010EA 2625 SEK
ACRO119150010 ACRO119150050
Fisetin
Fisetin
Formel: C₁₅H₁₀O₆
MW: 286.24 g/mol
Smältpunkt: >330 °C
MDL Number: MFCD00006829
CAS nummer: 528-48-3
EINECS: 208-434-4
Order Now
Specifikation testresultat
Appearance White to amber to dark green Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥90.0 %