AMMC iodide, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Varning
Synonyms: 3-[2-(N,N-diethyl-N-methylammonium)ethyl]-7-methoxy-4-methylcoumarin iodide
A1352-5MG
A1352-5MGEA 17280 SEK
A1352-5MG
AMMC iodide, Sigma-Aldrich®
AMMC iodide
AMMC is demethylated by Human CYP2D6 to the fluorescent product 3-[2-(N,N-diethyl-N-methylammonium)ethyl]-hydroxy-4-methylcoumarin.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b51ffe2a7a09367f8ea5b7dfd7b340ca31b897&vid=e0f9854337e644cdcb37d7e1e5fd6f83c43d9269&prid=web&clid=SAPDAM