AMMC iodide, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
|
Varning
|
Synonyms:
3-[2-(N,N-diethyl-N-methylammonium)ethyl]-7-methoxy-4-methylcoumarin iodide
A1352-5MG
A1352-5MGEA
17280
SEK
A1352-5MG
AMMC iodide, Sigma-Aldrich®
AMMC iodide
AMMC is demethylated by Human CYP2D6 to the fluorescent product 3-[2-(N,N-diethyl-N-methylammonium)ethyl]-hydroxy-4-methylcoumarin.