Ethyl eosin ≥95%, certified by the Biological Stain Commission, Sigma-Aldrich®
Leverantör: SIGMA-ALDRICH MICROSCOPY
Synonyms: 2',4',5',7'-Tetrabromoeosin ethyl ester potassium salt, Eosin alcohol soluble, Solvent Red 45
M-Clarity™ quality level = MQ200
199540-25G 199540-5G
199540-25GEA 3640 SEK
199540-25G 199540-5G
Ethyl eosin ≥95%, certified by the Biological Stain Commission, Sigma-Aldrich®
Ethyl eosin
Certified for use in the Stoval and Black method for demonstrating Negri bodies in the central nervous system of rabid animals.
Formel: C₂₂H₁₁Br₄KO₅
MW: 714,03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005039
CAS nummer: 6359-05-3
Order Now
Specifikation testresultat
λmax 532 nm