Brilliant cresyl blue, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Synonyms: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB
16030-10G 16030-50G
16030-10GEA 1050 SEK
16030-10G 16030-50G
Brilliant cresyl blue, Sigma-Aldrich®
Brilliant cresyl blue
'Brilliant Cresyl blue',for microscopy (Vit.), mixture of toluidine blue and waterblue ('Cresyl blue')
Formel: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00148901
CAS nummer: 81029-05-2