6-CFDA (6-Carboxyfluorescein diacetate, single isomer), Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Synonyms: 6-CFDA, 6-Carboxy-di-O-acetylfluorescein, 6-Carboxyfluorescein diacetate
C5041-100MG C5041-25MG
C5041-100MGEA 2940 SEK
C5041-100MG C5041-25MG
6-CFDA (6-Carboxyfluorescein diacetate, single isomer), Sigma-Aldrich®
6-CFDA (6-Carboxyfluorescein diacetate)
Used to differentiate viable cells from apoptotic cells. Live cells that are not apoptotic accumulate 6-carboxyfluorescein but do not accumulate annexin. Fluorescent wavelengths λex 492 nm; λemm 517 nm in 0.1 M Tris, pH 8.0.
Formel: C₂₅H₁₆O₉
MW: 460.39 g/mol
Storage Temperature: Frys
MDL Number: MFCD00036871
CAS nummer: 3348-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ade157b9d2dbb44f719042c0c830e5a8a3e75705&vid=26a78ce19f79b0a8e25ccf190e52ee99496f7d49&prid=web&clid=SAPDAM