Atorvastatin calcium salt trihydrate
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Calcium (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate trihydrate, Atorvastatin Calcium Salt Trihydrate, Atorvastatin hemicalcium salt sesquihydrate, Atorvastatin calcium
464810010 464810050
ACRO464810010EA 1600 SEK
ACRO464810010 ACRO464810050
Atorvastatin calcium salt trihydrate
Atorvastatin calcium salt trihydrate
  • Bioactive
Formel: C₆₆H₇₄CaF₂N₄O₁₃
MW: 1209.39 g/mol
Smältpunkt: 156 °C
Storage Temperature: Omgivande
MDL Number: MFCD09752074
CAS nummer: 344423-98-9
Order Now
Specifikation testresultat
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water 3.5 to 5.5 %
HPLC ≥97.5 %