Ambrisentan
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic acid, (S)-2-((4,6-Dimethylpyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid
463950010 463952500
ACRO463950010EA 7750 SEK
ACRO463950010 ACRO463952500
Ambrisentan
Ambrisentan
  • Bioactive
Formel: C₂₂H₂₂N₂O₄
MW: 378.42 g/mol
Smältpunkt: >150 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09842330
CAS nummer: 177036-94-1
Order Now
Specifikation testresultat
Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %