Astaxanthine (synthetic)
Leverantör: Thermo Scientific
Synonyms: Carophyll Pink, (3S,3'S)-Astaxanthin, Natupink, (6S,6'S)-3,3'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(6-hydroxy-2,4,4-trimethylcyclohex-2-enone), BioAstin
328610010 328612500
ACRO328610010EA 6200 SEK
ACRO328610010 ACRO328612500
Astaxanthine (synthetic)
Astaxanthine
  • Bioactive
Formel: C₄₀H₅₂O₄
MW: 596,852 g/mol
Smältpunkt: 216…218 °C
Storage Temperature: Frys
MDL Number: MFCD00672621
CAS nummer: 472-61-7
Order Now
Specifikation testresultat
Appearance Purplish-brown to purplish Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥96.0 %