Thymoquinone
Leverantör: Thermo Scientific
Varning
Synonyms: p-Mentha-3,6-diene-2,5-dione, 2-Isopropyl-5-methylcyclohexa-2,5-diene-1,4-dione, 5-Isopropyl-2-methyl-1,4-benzoquinone
305070010 305070050
ACRO305070010EA 890 SEK
ACRO305070010 ACRO305070050
Thymoquinone
Thymoquinone
  • Bioactive
Formel: C₁₀H₁₂O₂
MW: 164,20 g/mol
Kokpunkt: 230…232 °C (760 mmHg)
Smältpunkt: 43…44 °C
Flampunkt: 103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001602
CAS nummer: 490-91-5
Order Now
Specifikation testresultat
Appearance Yellow to orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 44 to 47 °C
GC ≥97.5 %