High visibility overalls, FR60
Leverantör: Portwest
FR60YNRM FR60YNRL FR60YNRS FR60YNRXXL FR60YNRXXXL FR60YNRXL FR60YNR4XL
PRTWFR60YNRMEA 3950 SEK
PRTWFR60YNRM PRTWFR60YNRL PRTWFR60YNRS PRTWFR60YNRXXL PRTWFR60YNRXXXL PRTWFR60YNRXL PRTWFR60YNR4XL
High visibility overalls, FR60
Workwear
FR60 is constructed with highly innovative Bizflame multi fabric. This comfortable garment offers protection against multiple risks including exposure to heat, fire, chemicals, electrical arcs and welding. Features include twin-stitched Hi-Vis strips on chest, arms, shoulders and legs, concealed two-way front brass zip, two concealed front chest pockets with piping detail, concealed side access pockets, hip pockets and rule pocket. The garment also has patented knee pad pockets designed with an innovative two tier adjustable system.
  • Protection against radiant, convective and contact heat
  • Certified protection against molten metal splash
  • Sew on flame resistant industrial wash tape
  • Two tier knee pad pockets allowing two positioning options
  • Radio loops for easy clipping of a radio
  • Triple-stitched seams for extra durability
  • 50+ UPF rated fabric to block 98% of UV rays
Guaranteed flame resistance for life of garment which is durable, strong and long lasting brass zip.
Certifikat: EN 1149 -5; EN 13034 Type 6; EN ISO 11612 (A1+A2, B1, C1, E3, F1); EN ISO 11611 Class 1 (A1+A2); IEC 61482-2 IEC 61482-1-2 Class 1; EN ISO 20471 Class 3; ASTM F1959/F1959M-12 (ATPV=13,6 Cal/cm² (HAF=82%)).