Methyllithium 1.6M i Dietyleter
Leverantör: Thermo Scientific
Fara
Synonyms: Methyllithium - lithium bromide complex, Methyllithium solution
Packaged under Nitrogen in resealable AcroSeal® bottles
H36516.9Z H36516.AE
H36516.9ZEA 4975 SEK
H36516.9Z H36516.AE
Methyllithium 1.6M i Dietyleter
Methyllithium
Formel: CH₃Li
MW: 21,975 g/mol
Flampunkt: -17°C (1°F)
Storage Temperature: Omgivande
MDL Number: MFCD00008253
CAS nummer: 917-54-4
EINECS: 213-026-4
UN: 3394
ADR: 4.2,I