Pyronin G, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
M-Clarity™ quality level = MQ200
213519-1G 213519-5G
213519-1GEA 713 SEK
213519-1G 213519-5G
Pyronin G, Sigma-Aldrich®
Pyronin G
Formel: C₁₇H₁₉ClN₂O
MW: 302.8 g/mol
Smältpunkt: 250…260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011725
CAS nummer: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6029405bdb971ff6871b35ad2342de4b97699ad&vid=83111b2c1d0c86c1135e9b11d0342467198b3dd9&prid=web&clid=SAPDAM