Pyronin G, certified
Leverantör: Thermo Scientific
Varning
Synonyms: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
418630010 418630050
ACRO418630010EA 775 SEK
ACRO418630010 ACRO418630050
Pyronin G, certified
Pyronine G
Formel: C₁₇H₁₉ClN₂O
MW: 302,8 g/mol
Smältpunkt: 250…260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011725
CAS nummer: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190
Order Now
Specifikation testresultat
Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥45 %
UV Lambda max 546 to 551 nm
Absorption Ratio P-15/P+15 1.23 to 1.7
BSC Certification Passes