1,3,5-Triazine 97
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: s-Triazine
L16911.03 L16911.06 L16911.14
L16911.03EA 1175 SEK
L16911.03 L16911.06 L16911.14
1,3,5-Triazine 97
1,3,5-Triazine
Purity: 97% CAS No.: 290-87-9 Molecular Formula: C3H3N3 Formula Weight: 81.08 Melting Point: 77-83° dec. Boiling Point: 114° MDL No.: MFCD00006044
Formel: C₃H₃N₃
MW: 81.08 g/mol
Kokpunkt: 114 °C
Smältpunkt: 77…83 °C (dec.)
Densitet: 1.380
Storage Temperature: Omgivande
MDL Number: MFCD00006044
CAS nummer: 290-87-9
EINECS: 206-028-1
UN: 2928
ADR: 6.1 II