1,4-Butanesultone ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,4-Butane sultone
L03992.14 L03992.22
L03992.14EA 1025 SEK
L03992.14 L03992.22
1,4-Butanesultone ≥99%
1,4-Butanesultone
Purity: 99% CAS No.: 1633-83-6 Molecular Formula: C4H8O3S Formula Weight: 136.17 Flash Point: 153°(307°F) Refractive Index: 1.4640 Melting Point: 12-15° Boiling Point: 114-116°/1.5mm Beilstein Reference: 110588 MDL No.: MFCD00006584
Formel: C₄H₈O₃S
MW: 136,17 g/mol
Kokpunkt: 114…116 °C (1,5 mmHg)
Smältpunkt: 12…15 °C
Densitet: 1,335
Flampunkt: 153 °C (307 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006584
CAS nummer: 1633-83-6
EINECS: 216-647-9