Tetrahydro-4H-pyran-4-one
Leverantör: Thermo Scientific
Synonyms:
Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one
SB01650DA
SB01650EA
MAYBSB01650DAEA
0
SEK
MAYBSB01650DA
MAYBSB01650EA
Tetrahydro-4H-pyran-4-one
Tetrahydro-4H-pyran-4-one
Formel:
C₅H₈O₂ MW: 100,12 g/mol Kokpunkt: 166 °C (1013 hPa) Densitet: 1,08 g/cm³ (20 °C) |
MDL Number:
MFCD00006581 CAS nummer: 29943-42-8 EINECS: 249-967-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...