Chemical resistant overalls, AlphaTec® 3000, Model 111
Leverantör: Ansell
YE30W-00111-08 YE30W-00111-09
114-9509EA 0 SEK
114-9509 114-9510
Chemical resistant overalls, AlphaTec® 3000, Model 111
Engångskläder
Durable, comfortable multi-layer chemical barrier, offering Type 3/4/5 protection.
  • Multi-layer barrier fabric provides effective protection against numerous chemicals
  • Lightweight and durable
  • Highly visible bright yellow color for improved worker safety
  • Anti-static, tested according to EN 1149-5 and AATCC 76
Coveralls are designed to protect, with innovative features that include a 2-piece hood, double zip closure, and double cuffs. Elasticated hood, waist, double cuffs, and ankles. Garments are manufactured from multi-layer, non-woven barrier laminate fabric with ultrasonically welded and taped seams.
Recommended for transferring liquids and solids between vessels, tanks, and process equipment, unexpected leakages or spills, opening and draining pumps valves or lines, loading and unloading trucks and vehicles, filling, blending and charging of raw materials, product inspection, mixing or loading chemicals, and plant and machine cleaning.
Certifikat: Type 3-B; Type 4-B; Type 5-B; EN 1073-2; EN 14126; EN 1149-5; Category III
Förpackning: Packaged 15 pieces/carton.