Maltol 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
121550250 121551000 121550050 121555000
ACRO121550250EA 785 SEK
ACRO121550250 ACRO121551000 ACRO121550050 ACRO121555000
Maltol 99%
Maltol
Formel: C₆H₆O₃
MW: 126,11 g/mol
Kokpunkt: 170 °C (100 mmHg)
Smältpunkt: 160…164 °C
Densitet: 1,453 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006578
CAS nummer: 118-71-8
EINECS: 204-271-8
Merck Index: 13,05735
Order Now
Specifikation testresultat
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
Water ≤0.5 %
GC ≥98.5 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
UV at 240 nm A: ≤0.05
Color scale ≤10 APHA
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g