Iron(III) acetylacetonate 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')
119130250 119131000 119132500 119130010
ACRO119130250EA 490 SEK
ACRO119130250 ACRO119131000 ACRO119132500 ACRO119130010
Iron(III) acetylacetonate 99+%
Iron(III) acetylacetonate
Formel: C₁₅H₂₁FeO₆
MW: 353,18 g/mol
Kokpunkt: 100 °C (9,7513 -rr)
Smältpunkt: 180…183 °C (dec.)
Densitet: 5,24 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000020
CAS nummer: 14024-18-1
EINECS: 237-853-5
Order Now
Specifikation testresultat
Appearance Dark orange to dark red Powder or crystals
Infrared spectrum Conforms
Titration Iodometric ≥99.0 %
HPLC ≥94 %