Molybdentrioxid 99.5%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Molybdenum trioxide
A11159.0B A11159.18 A11159.30
A11159.0BEA 6150 SEK
A11159.0B A11159.18 A11159.30
Molybdentrioxid 99.5%
Molybdentrioxid
Purity: 99.5% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Odour: Odourless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469
Formel: MoO₃
Kokpunkt: 1155 °C (subl.)
Smältpunkt: 795 °C
Densitet: 4,692
Storage Temperature: Omgivande
MDL Number: MFCD00003469
CAS nummer: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,6239