Molybdentrioxid ≥99.9995%, Puratronic®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Molybdenum trioxide
12930.22 12930.14 12930.36
12930.22EA 13500 SEK
12930.22 12930.14 12930.36
Molybdentrioxid ≥99.9995%, Puratronic®
Molybdentrioxid
Purity: 99.9995% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Physical Form: Powder Density: 4.692 Appearance: White to grey Odour: Odourless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469
Formel: MoO₃
MW: 143.94 g/mol
Kokpunkt: 1155 °C (subl.)
Smältpunkt: 795
Densitet: 4.692
Storage Temperature: Omgivande
MDL Number: MFCD00003469
CAS nummer: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1 III
Merck Index: 14,6239