Molybdenum pentachloride 95%
Leverantör: Thermo Scientific
Fara
197080100 197081000
ACRO197080100EA 1025 SEK
ACRO197080100 ACRO197081000
Molybdenum pentachloride 95%
Molybdenum pentachloride
Formel: Cl₅Mo
MW: 273,2 g/mol
Kokpunkt: 268 °C (1013 hPa)
Smältpunkt: 194 °C
Densitet: 2,928 g/cm³ (20 °C)
MDL Number: MFCD00003467
CAS nummer: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Blue to green-black to black Crystalline powder or crystals
Titration Argentometric ≥94 %