Zinkoxid substrate 10x10x0.5mm, polished one side 0001 orientation
Leverantör: Thermo Scientific
Varning
Synonyms: Zinci oxidum, Zinc (II) oxide
45486.KT 45486.KU
45486.KTEA 20075 SEK
45486.KT 45486.KU
Zinkoxid substrate 10x10x0.5mm, polished one side 0001 orientation
Zinkoxid
CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Odour: Odourless MDL No.: MFCD00011109
Formel: ZnO
MW: 81,39 g/mol
Smältpunkt: 1975 °C
Densitet: 5,61 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-171-9
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200