Silverkromat 99,9 Metaller basis
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Argentous chromate, Silver(I) chromate
45495.06 45495.22
45495.06EA 980 SEK
45495.06 45495.22
Silverkromat 99,9 Metaller basis
Silverkromat
Purity: 99.9% CAS No.: 7784-01-2 Molecular Formula: O4Ag2Cr Formula Weight: 331.73 Physical Form: Powder Density: 5.63 Appearance: Red-brown Odour: Odourless Merck Reference: 14,8510 UN No.: 1479 MDL No.: MFCD00003402
Formel: Ag₂CrO₄
MW: 331.73 g/mol
Kokpunkt: 1550
Densitet: 5.63
Storage Temperature: Omgivande
MDL Number: MFCD00003402
CAS nummer: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1 III
Merck Index: 14,8510